Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEFF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305134.1 | NP_001292063.1 | 1, 2 | 585 | ||
NM_001305145.1 | NP_001292074.1 | 1, 2 | 585 | |||
NM_016192.3 | NP_057276.2 | 1, 2 | 585 | |||
XM_011510890.2 | XP_011509192.1 | 1, 2 | 580 | |||
XM_017003739.1 | XP_016859228.1 | 1, 2 | 592 | |||
XM_017003740.1 | XP_016859229.1 | 1, 2 | 592 | |||
GenBank mRNA | AB004064.1 | - | 254 | |||
AB017269.1 | - | 1, 2 | 543 | |||
AF179274.2 | - | 1, 2 | 570 | |||
AF242222.1 | - | 1, 2 | 585 | |||
AK054646.1 | - | 1, 2 | 1365 | |||
AK074507.1 | - | 1, 2 | 199 | |||
AK074632.1 | - | 1, 2 | 568 | |||
AK222691.1 | - | 1, 2 | 590 | |||
AK315376.1 | - | 1, 2 | 586 | |||
AL157430.1 | - | 1, 2 | 568 | |||
AY358907.1 | - | 1, 2 | 551 | |||
BC008973.1 | - | 1, 2 | 526 | 4180709 | ||
CR457390.1 | - | 1, 2 | 176 | |||
DQ133599.1 | - | 1, 2 | 176 |