Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PISD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326411.1 | NP_001313340.1 | 8 | 1422 | ||
NM_001326412.1 | NP_001313341.1 | 8 | 1359 | |||
NM_001326413.1 | NP_001313342.1 | 8 | 1508 | |||
NM_001326414.1 | NP_001313343.1 | 9 | 1588 | |||
NM_001326415.1 | NP_001313344.1 | 9 | 1762 | |||
NM_001326416.1 | NP_001313345.1 | 11 | 1951 | |||
NM_001326417.1 | NP_001313346.1 | 10 | 1842 | |||
NM_001326418.1 | NP_001313347.1 | 7 | 1342 | |||
NM_001326419.1 | NP_001313348.1 | 10 | 1728 | |||
NM_001326420.1 | NP_001313349.1 | 7 | 1228 | |||
NM_001326421.1 | NP_001313350.1 | 7 | 1261 | |||
NM_014338.3 | NP_055153.1 | 9 | 1394 | |||
NM_178022.1 | NP_821141.1 | 8 | 1303 | |||
GenBank mRNA | AF035304.1 | - | 1 | 72 | ||
AF086277.1 | - | 2 | 79 | |||
AL050371.1 | - | 7 | 1249 | |||
BC001482.2 | - | 9 | 1394 | 3508281 | ||
BC009315.1 | - | 3 | 269 | |||
CR456540.1 | - | 8 | 1072 |