Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013231.4 | NP_037363.1 | 2 | 1413 | ||
XM_005267490.3 | XP_005267547.1 | 2 | 1512 | |||
XM_011536611.2 | XP_011534913.1 | 2 | 1153 | |||
XM_017021129.1 | XP_016876618.1 | 2 | 1694 | |||
XM_017021130.1 | XP_016876619.1 | 2 | 10968 | |||
XM_017021131.1 | XP_016876620.1 | 3 | 10122 | |||
XM_017021132.1 | XP_016876621.1 | 3 | 1601 | |||
XM_017021133.1 | XP_016876622.1 | 3 | 1619 | |||
XM_017021134.1 | XP_016876623.1 | 3 | 1546 | |||
XM_017021135.1 | XP_016876624.1 | 2 | 1414 | |||
GenBank mRNA | AF169676.1 | - | 1 | 703 | ||
AK056649.1 | - | 2 | 1413 | |||
AY358287.1 | - | 1 | 909 | |||
BC126249.1 | - | 2 | 1329 | 8991965 | ||
BC130290.1 | - | 2 | 1329 | 40146321 | ||
BC143936.1 | - | 2 | 1351 | 9052454 |