Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FKBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308373.1 | NP_001295302.1 | 3 | 442 | ||
NM_012181.4 | NP_036313.3 | 3 | 442 | |||
XM_011527863.1 | XP_011526165.1 | 3 | 500 | |||
XM_011527864.1 | XP_011526166.1 | 2 | 331 | |||
GenBank mRNA | AK026952.1 | - | 1030 | |||
AK300571.1 | - | 3 | 416 | |||
AK313363.1 | - | 3 | 373 | |||
AY225339.1 | - | 2 | 306 | |||
AY278607.1 | - | 3 | 369 | |||
BC009966.1 | - | 2 | 149 | 4299915 | ||
BX538124.1 | - | 3 | 420 | |||
BX647405.1 | - | 3 | 419 | |||
BX647720.1 | - | 3 | 377 |