Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001752.3 | NP_001743.1 | 3 | 386 | ||
GenBank mRNA | AK291585.1 | - | 3 | 386 | ||
AK301577.1 | - | 2 | 156 | |||
AK310184.1 | - | 2 | 211 | |||
AY028632.1 | - | 3 | 297 | |||
BC110398.1 | - | 3 | 375 | 5551309 | ||
BC112217.1 | - | 3 | 335 | 8327685 | ||
BC112219.1 | - | 3 | 335 | 8327687 | ||
X04076.1 | - | 3 | 367 |