Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014345.2 | NP_055160.2 | 3 | 651 | ||
XM_011514451.2 | XP_011512753.1 | 3 | 252 | |||
XM_011514452.2 | XP_011512754.1 | 3 | 884 | |||
GenBank mRNA | AF090114.1 | - | 3 | 654 | ||
AF121141.1 | - | 3 | 389 | |||
BC030687.1 | - | 3 | 686 | |||
BC098434.1 | - | 3 | 626 |