Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD54B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205263.1 | NP_001192192.1 | 5 | 1297 | ||
NM_012415.3 | NP_036547.1 | 7 | 1274 | |||
GenBank mRNA | AF112481.1 | - | 7 | 1223 | ||
AK290437.1 | - | 7 | 1244 | |||
AK307516.1 | - | 5 | 970 | |||
AL133578.1 | - | 1 | 152 | |||
BC001965.2 | - | 7 | 1216 | 3452509 | ||
BC020668.1 | - | 8 | 1287 |