Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015380.4 | NP_056195.3 | 6 | 646 | ||
GenBank mRNA | AF151809.1 | - | 6 | 558 | ||
AK001087.1 | - | 6 | 585 | |||
AK222657.1 | - | 6 | 520 | |||
AK312773.1 | - | 6 | 481 | |||
AY189688.1 | - | 6 | 561 | |||
BC007830.1 | - | 6 | 553 | 3635175 | ||
BC011681.2 | - | 6 | 606 | 3010765 | ||
BC015200.1 | - | 6 | 496 | 4333051 | ||
CR456483.1 | - | 6 | 458 |