Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242374.1 | NP_001229303.1 | 3 | 1166 | ||
NM_001242375.1 | NP_001229304.1 | 3 | 751 | |||
NM_015391.3 | NP_056206.1 | 3 | 532 | |||
GenBank mRNA | AK001285.1 | - | 3 | 1163 | ||
AK026550.1 | - | 3 | 486 | |||
AL050051.1 | - | 3 | 751 | |||
AV682398.1 | - | 3 | 497 | |||
BC005398.1 | - | 3 | 498 | 3828623 | ||
BU852706.1 | - | 3 | 511 | |||
BX537696.1 | - | 4 | 952 |