Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RUNX1T1 SIRNA Assays ›
Gene Symbol:
RUNX1T1
Gene Name:
RUNX1 translocation partner 1
Gene Aliases:
AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2
Chromosome Location:
Chr.8: 91954967 - 92103365 on Build GRCh38
UniGene ID:
Hs.368431
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2462
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RUNX1T1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198625.1
NP_001185554.1
10
1573
NM_001198626.1
NP_001185555.1
8
1431
NM_001198627.1
NP_001185556.1
9
1491
NM_001198628.1
NP_001185557.1
7
1088
NM_001198629.1
NP_001185558.1
7
1214
NM_001198630.1
NP_001185559.1
8
1274
NM_001198631.1
NP_001185560.1
7
1287
NM_001198632.1
NP_001185561.1
7
1205
NM_001198633.1
NP_001185562.1
6
1176
NM_001198634.1
NP_001185563.1
6
1114
NM_001198679.1
NP_001185608.1
7
1258
NM_004349.3
NP_004340.1
6
1224
NM_175634.2
NP_783552.1
7
1349
NM_175635.2
NP_783553.1
6
1041
NM_175636.2
NP_783554.1
5
1123
XM_006716676.3
XP_006716739.1
8
1158
XM_011517351.2
XP_011515653.1
8
1203
XM_011517352.2
XP_011515654.1
8
1443
XM_011517353.2
XP_011515655.1
7
1151
XM_017013930.1
XP_016869419.1
7
1459
XM_017013931.1
XP_016869420.1
6
981
XM_017013932.1
XP_016869421.1
8
1346
XM_017013933.1
XP_016869422.1
9
1410
XM_017013934.1
XP_016869423.1
9
1518
XM_017013935.1
XP_016869424.1
8
1323
XM_017013936.1
XP_016869425.1
7
1417
XM_017013937.1
XP_016869426.1
8
1243
XM_017013938.1
XP_016869427.1
7
1107
XM_017013939.1
XP_016869428.1
2
874
XM_017013940.1
XP_016869429.1
3
2103
XM_017013941.1
XP_016869430.1
2
1893
GenBank mRNA
AK297616.1
-
6
938
AK312592.1
-
10
1335
BC005850.1
-
6
1003
2961112
BC067078.1
-
6
994
2964656
BC139783.1
-
2
1816
BT009871.1
-
5
783
CR456792.1
-
5
813
D14289.1
-
5
1080
D14821.1
-
6
978
D43638.1
-
6
1224
KF946506.1
-
4
374
KF946507.1
-
6
842
KF946508.1
-
6
842
KF946511.1
-
6
828
KF946512.1
-
7
2132
KF946513.1
-
6
2512
KF946514.1
-
6
828
KF946516.1
-
5
1068
KF946517.1
-
5
1068
KF946518.1
-
5
1068
KF946520.1
-
2
490
KF946521.1
-
2
490
KF946522.1
-
2
490
KF946523.1
-
2
490
X79990.1
-
5
806
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108657, XM_006237903, XM_008763549, XM_006237904, XM_008763548, and others.
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
generation of precursor metabolites and energy
transcription, DNA-templated
negative regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline