Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM149A SIRNA Assays ›
Gene Symbol:
FAM149A
Gene Name:
family with sequence similarity 149 member A
Gene Aliases:
MST119, MSTP119
Chromosome Location:
Chr.4: 186104798 - 186172663 on Build GRCh38
UniGene ID:
Hs.744961
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24629
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM149A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006655.2
NP_001006656.1
11
1510
NM_015398.2
NP_056213.1
12
1809
XM_005262911.1
XP_005262968.1
12
1770
XM_005262912.1
XP_005262969.1
12
1841
XM_005262913.1
XP_005262970.1
12
1678
XM_011531834.1
XP_011530136.1
11
1537
XM_011531835.2
XP_011530137.1
12
1770
XM_011531836.1
XP_011530138.1
11
1537
XM_017007996.1
XP_016863485.1
12
2355
XM_017007997.1
XP_016863486.1
12
2352
XM_017007998.1
XP_016863487.1
11
1537
XM_017007999.1
XP_016863488.1
12
3142
XM_017008000.1
XP_016863489.1
12
1626
XM_017008001.1
XP_016863490.1
12
1715
XM_017008002.1
XP_016863491.1
12
1770
XM_017008003.1
XP_016863492.1
12
1626
XM_017008004.1
XP_016863493.1
12
1678
XM_017008005.1
XP_016863494.1
12
1783
XM_017008006.1
XP_016863495.1
12
1678
XM_017008007.1
XP_016863496.1
12
1841
GenBank mRNA
AF173894.1
-
11
1446
AK057166.1
-
12
1639
AK297679.1
-
12
1749
AL080065.1
-
6
858
BC110816.1
-
12
1711
BC136792.1
-
9
1255
9020782
BC142992.1
-
12
1763
8860446
BC144411.1
-
9
1255
9052932
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-8tvtn:80/10.239.165.222:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0