Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079880.1 | NP_001073349.1 | 15 | 2393 | ||
NM_001079881.1 | NP_001073350.1 | 15 | 2257 | |||
NM_001079882.1 | NP_001073351.1 | 13 | 1994 | |||
NM_016457.4 | NP_057541.2 | 14 | 2672 | |||
XM_005258716.2 | XP_005258773.2 | 13 | 1650 | |||
GenBank mRNA | AF151021.1 | - | 2 | 196 | ||
AF309082.1 | - | 14 | 1954 | |||
AK074673.1 | - | 13 | 1633 | |||
AK075266.1 | - | 13 | 1743 | |||
AK092896.1 | - | 14 | 2044 | |||
AK095884.1 | - | 15 | 1936 | |||
AK300339.1 | - | 15 | 2207 | |||
AK303629.1 | - | 13 | 1648 | |||
AL050147.1 | - | 8 | 909 | |||
BC015472.1 | - | 5 | 371 | |||
BC025307.1 | - | 10 | 1243 |