Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278473.2 | NP_001265402.1 | 4 | 846 | ||
NM_001304390.1 | NP_001291319.1 | 3 | 738 | |||
NM_001304391.1 | NP_001291320.1 | 3 | 738 | |||
NM_001304415.1 | NP_001291344.1 | 3 | 738 | |||
NM_001304416.1 | NP_001291345.1 | 2 | 644 | |||
NM_001304417.1 | NP_001291346.1 | 1 | 436 | |||
NM_015424.5 | NP_056239.3 | 4 | 846 | |||
GenBank mRNA | AF332891.1 | - | 2 | 121 | ||
AK304519.1 | - | 4 | 614 | |||
AY163868.1 | - | 4 | 352 | |||
AY279090.1 | - | 4 | 643 | |||
AY279091.1 | - | 4 | 643 | |||
AY279092.1 | - | 2 | 644 | |||
AY279093.1 | - | 3 | 738 | |||
AY279094.1 | - | 2 | 644 | |||
AY358522.1 | - | 4 | 635 | |||
BC142623.1 | - | 4 | 846 | 40147467 | ||
JN579714.1 | - | 4 | 349 |