Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABI3BP SIRNA Assays ›
Gene Symbol:
ABI3BP
Gene Name:
ABI family member 3 binding protein
Gene Aliases:
NESHBP, TARSH
Chromosome Location:
Chr.3: 100749335 - 100993508 on Build GRCh38
UniGene ID:
Hs.477015
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24683
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABI3BP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015429.3
NP_056244.2
23
2030
XM_005247281.2
XP_005247338.1
55
4112
XM_005247282.2
XP_005247339.1
54
4070
XM_005247283.2
XP_005247340.1
54
4064
XM_005247284.2
XP_005247341.1
54
4055
XM_005247285.2
XP_005247342.1
54
4052
XM_005247287.2
XP_005247344.1
54
4046
XM_005247288.2
XP_005247345.1
54
4040
XM_005247289.2
XP_005247346.1
54
4037
XM_005247290.2
XP_005247347.1
55
4112
XM_005247293.2
XP_005247350.1
53
4004
XM_005247294.2
XP_005247351.1
53
3995
XM_005247295.2
XP_005247352.1
53
3974
XM_005247296.2
XP_005247353.1
53
3971
XM_005247298.2
XP_005247355.1
53
3968
XM_005247299.3
XP_005247356.1
53
3980
XM_005247300.2
XP_005247357.1
52
3938
XM_005247302.2
XP_005247359.1
52
3908
XM_005247303.2
XP_005247360.1
51
3851
XM_005247305.2
XP_005247362.1
50
3767
XM_005247306.2
XP_005247363.1
48
3704
XM_005247309.1
XP_005247366.1
53
3745
XM_006713570.2
XP_006713633.1
52
3926
XM_006713571.2
XP_006713634.1
52
3896
XM_006713572.2
XP_006713635.1
52
3890
XM_011512639.1
XP_011510941.1
54
4052
XM_011512640.1
XP_011510942.1
54
4046
XM_011512641.1
XP_011510943.1
54
4037
XM_011512643.1
XP_011510945.1
53
4031
XM_011512644.1
XP_011510946.1
54
4028
XM_011512645.1
XP_011510947.1
54
4028
XM_011512646.1
XP_011510948.1
54
4019
XM_011512647.1
XP_011510949.1
53
3977
XM_011512648.1
XP_011510950.1
53
3974
XM_011512649.1
XP_011510951.1
53
3968
XM_011512651.1
XP_011510953.1
52
3911
XM_011512652.1
XP_011510954.1
52
3899
XM_011512653.1
XP_011510955.1
51
3890
XM_011512654.1
XP_011510956.1
52
3881
XM_011512656.1
XP_011510958.1
51
3815
XM_011512657.1
XP_011510959.1
52
3887
XM_011512658.1
XP_011510960.1
50
3800
XM_011512659.1
XP_011510961.1
50
3779
XM_011512660.1
XP_011510962.1
50
3749
XM_017006105.1
XP_016861594.1
54
4055
XM_017006106.1
XP_016861595.1
54
4037
XM_017006107.1
XP_016861596.1
53
3980
XM_017006108.1
XP_016861597.1
50
3815
GenBank mRNA
AB056106.1
-
5
466
AK093861.1
-
5
289
AK123737.1
-
15
1057
AK123748.1
-
13
846
AK299938.1
-
33
2112
BC030221.1
-
23
2010
5205716
BX648726.1
-
31
2119
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cell-substrate adhesion
extracellular matrix organization
collagen binding
heparin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0