Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001755.2 | NP_001746.1 | 3 | 515 | ||
NM_022845.2 | NP_074036.1 | 3 | 515 | |||
XM_005256212.3 | XP_005256269.1 | 3 | 477 | |||
XM_017023813.1 | XP_016879302.1 | 3 | 501 | |||
GenBank mRNA | AA020956.1 | - | 3 | 320 | ||
AF294326.1 | - | 3 | 254 | |||
AK290462.1 | - | 3 | 273 | |||
AK291834.1 | - | 3 | 257 | |||
BC018509.1 | - | 3 | 515 | 3867061 | ||
BT006862.1 | - | 3 | 254 | |||
L20298.1 | - | 3 | 233 |