Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199069.1 | NP_951032.1 | 3 | 791 | ||
NM_199070.1 | NP_951033.1 | 3 | 354 | |||
NM_199073.1 | NP_951047.1 | 3 | 425 | |||
NM_199074.1 | NP_951056.1 | 3 | 306 | |||
GenBank mRNA | AL049955.1 | - | 3 | 358 | ||
BC002873.2 | - | 3 | 341 | 3945015 | ||
BC111004.1 | - | 3 | 1421 | 5744973 | ||
BM354128.1 | - | 2 | 397 | |||
BQ652614.1 | - | 3 | 306 | |||
CD358942.1 | - | 3 | 495 |