Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015483.2 | NP_056298.2 | 4 | 2362 | ||
XM_005249696.2 | XP_005249753.1 | 4 | 2218 | |||
XM_005249697.2 | XP_005249754.1 | 4 | 1951 | |||
XM_011515256.2 | XP_011513558.1 | 4 | 2097 | |||
XM_011515257.2 | XP_011513559.1 | 4 | 1831 | |||
GenBank mRNA | AB040922.1 | - | 1 | 2986 | ||
AK024557.1 | - | 1 | 1327 | |||
AK292802.1 | - | 4 | 2084 | |||
AL117562.1 | - | 1 | 880 | |||
BC037887.2 | - | 4 | 2266 | 5274706 | ||
BC047107.1 | - | 4 | 2099 | 5275217 |