Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283014.1 | NP_001269943.1 | 3 | 389 | ||
NM_001283015.1 | NP_001269944.1 | 3 | 389 | |||
NM_001283016.1 | NP_001269945.1 | 4 | 461 | |||
NM_001283017.1 | NP_001269946.1 | 4 | 461 | |||
NM_001283019.1 | NP_001269948.1 | 3 | 389 | |||
NM_015901.5 | NP_056985.3 | 3 | 389 | |||
NR_104264.1 | 3 | 389 | ||||
XM_017016033.1 | XP_016871522.1 | 3 | 309 | |||
GenBank mRNA | AK300980.1 | - | 4 | 314 | ||
AK303235.1 | - | 3 | 265 | |||
AK309755.1 | - | 3 | 242 | |||
AK316538.1 | - | 4 | 314 | |||
AL521667.3 | - | 3 | 232 | |||
BC031224.1 | - | 3 | 309 | 4824935 | ||
BC046173.2 | - | 3 | 242 | 3873776 |