Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2CD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015500.1 | NP_056315.1 | 10 | 1405 | ||
NM_199050.2 | NP_950251.1 | 9 | 904 | |||
XM_005261109.4 | XP_005261166.1 | 9 | 945 | |||
XM_011529522.1 | XP_011527824.1 | 10 | 1505 | |||
XM_011529523.2 | XP_011527825.1 | 11 | 2171 | |||
XM_011529524.2 | XP_011527826.1 | 10 | 1505 | |||
XM_017028315.1 | XP_016883804.1 | 7 | 641 | |||
GenBank mRNA | AB047784.1 | - | 10 | 1405 | ||
AB047785.1 | - | 10 | 1405 | |||
AK128550.1 | - | 7 | 744 | |||
AL833323.1 | - | 9 | 904 | |||
BC062323.1 | - | 9 | 886 | 5419622 | ||
CR627455.1 | - | 2 | 1179 |