Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIGD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099668.1 | NP_001093138.1 | 2 | 340 | ||
NM_001099669.1 | NP_001093139.1 | 2 | 193 | |||
NM_014056.3 | NP_054775.2 | 2 | 169 | |||
GenBank mRNA | AF077034.1 | - | 2 | 121 | ||
AF145385.1 | - | 2 | 108 | |||
AK225461.1 | - | 2 | 93 | |||
AK225571.1 | - | 2 | 124 | |||
AK311815.1 | - | 2 | 124 | |||
AL110233.1 | - | 2 | 139 | |||
BC000601.1 | - | 2 | 107 | 3346355 | ||
BC009583.1 | - | 2 | 100 | 4093421 | ||
BC009594.1 | - | 2 | 106 | 4109190 | ||
BC070277.1 | - | 2 | 122 | 4686431 | ||
BG708126.1 | - | 2 | 174 | |||
BP208784.1 | - | 2 | 339 | |||
BP370118.1 | - | 2 | 115 | |||
CR457385.1 | - | 1 | 47 | |||
DA428926.1 | - | 2 | 335 |