Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SIPA1L1 SIRNA Assays ›
Gene Symbol:
SIPA1L1
Gene Name:
signal induced proliferation associated 1 like 1
Gene Aliases:
E6TP1
Chromosome Location:
Chr.14: 71320418 - 71741229 on Build GRCh38
UniGene ID:
Hs.654657
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24984
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SIPA1L1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284245.1
NP_001271174.1
10
3523
NM_001284246.1
NP_001271175.1
10
4112
NM_001284247.1
NP_001271176.1
9
3464
NM_015556.2
NP_056371.1
10
3523
XM_005267516.4
XP_005267573.1
10
4112
XM_006720111.2
XP_006720174.1
12
5402
XM_011536630.1
XP_011534932.1
14
3874
XM_011536631.1
XP_011534933.1
14
4285
XM_011536632.1
XP_011534934.1
13
3794
XM_011536635.2
XP_011534937.1
13
3941
XM_011536638.2
XP_011534940.1
9
2597
XM_017021159.1
XP_016876648.1
16
4031
XM_017021160.1
XP_016876649.1
14
3953
XM_017021161.1
XP_016876650.1
15
3906
XM_017021162.1
XP_016876651.1
17
4088
XM_017021163.1
XP_016876652.1
12
3755
XM_017021164.1
XP_016876653.1
12
3762
XM_017021165.1
XP_016876654.1
14
3831
XM_017021166.1
XP_016876655.1
15
3964
XM_017021167.1
XP_016876656.1
14
3839
XM_017021168.1
XP_016876657.1
14
3813
XM_017021169.1
XP_016876658.1
13
3966
XM_017021170.1
XP_016876659.1
13
4324
XM_017021171.1
XP_016876660.1
14
3897
XM_017021172.1
XP_016876661.1
13
3772
XM_017021173.1
XP_016876662.1
12
3899
XM_017021174.1
XP_016876663.1
12
4139
XM_017021175.1
XP_016876664.1
12
4257
XM_017021176.1
XP_016876665.1
17
4084
XM_017021177.1
XP_016876666.1
17
4073
XM_017021178.1
XP_016876667.1
15
3911
XM_017021179.1
XP_016876668.1
14
3814
XM_017021180.1
XP_016876669.1
18
4126
XM_017021181.1
XP_016876670.1
16
4031
XM_017021182.1
XP_016876671.1
15
3964
XM_017021183.1
XP_016876672.1
13
3772
XM_017021184.1
XP_016876673.1
17
4084
XM_017021185.1
XP_016876674.1
10
4112
XM_017021186.1
XP_016876675.1
16
4031
XM_017021187.1
XP_016876676.1
14
3897
XM_017021188.1
XP_016876677.1
13
3772
XM_017021189.1
XP_016876678.1
15
3881
XM_017021190.1
XP_016876679.1
16
4031
XM_017021191.1
XP_016876680.1
14
3839
XM_017021192.1
XP_016876681.1
13
3772
XM_017021193.1
XP_016876682.1
13
3770
XM_017021194.1
XP_016876683.1
16
4031
XM_017021195.1
XP_016876684.1
16
4031
XM_017021196.1
XP_016876685.1
16
4031
XM_017021197.1
XP_016876686.1
15
2231
XM_017021198.1
XP_016876687.1
12
1972
XM_017021199.1
XP_016876688.1
16
2273
XM_017021200.1
XP_016876689.1
13
2039
GenBank mRNA
AB007900.2
-
9
3247
AF090989.1
-
10
3510
AF090990.1
-
10
3510
AK122930.1
-
10
4112
AK302650.1
-
9
1762
BC146777.1
-
9
3247
9007226
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
actin cytoskeleton reorganization
regulation of GTPase activity
ephrin receptor signaling pathway
regulation of synaptic plasticity
regulation of axonogenesis
regulation of small GTPase mediated signal transduction
regulation of dendritic spine morphogenesis
activation of GTPase activity
molecular_function
GTPase activator activity
ephrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0