Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SIPA1L1 SIRNA Assays ›
Gene Symbol:
SIPA1L1
Gene Name:
signal induced proliferation associated 1 like 1
Gene Aliases:
E6TP1
Chromosome Location:
Chr.14: 71320418 - 71741229 on Build GRCh38
UniGene ID:
Hs.654657
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24986
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SIPA1L1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284245.1
NP_001271174.1
7
2484
NM_001284246.1
NP_001271175.1
7
3073
NM_001284247.1
NP_001271176.1
6
2425
NM_015556.2
NP_056371.1
7
2484
XM_005267516.4
XP_005267573.1
7
3073
XM_006720111.2
XP_006720174.1
9
4363
XM_011536630.1
XP_011534932.1
11
2835
XM_011536631.1
XP_011534933.1
11
3246
XM_011536632.1
XP_011534934.1
10
2755
XM_011536635.2
XP_011534937.1
10
2902
XM_011536638.2
XP_011534940.1
6
1558
XM_017021159.1
XP_016876648.1
13
2992
XM_017021160.1
XP_016876649.1
11
2914
XM_017021161.1
XP_016876650.1
12
2867
XM_017021162.1
XP_016876651.1
14
3049
XM_017021163.1
XP_016876652.1
9
2716
XM_017021164.1
XP_016876653.1
9
2723
XM_017021165.1
XP_016876654.1
11
2792
XM_017021166.1
XP_016876655.1
12
2925
XM_017021167.1
XP_016876656.1
11
2800
XM_017021168.1
XP_016876657.1
11
2774
XM_017021169.1
XP_016876658.1
10
2927
XM_017021170.1
XP_016876659.1
10
3285
XM_017021171.1
XP_016876660.1
11
2858
XM_017021172.1
XP_016876661.1
10
2733
XM_017021173.1
XP_016876662.1
9
2860
XM_017021174.1
XP_016876663.1
9
3100
XM_017021175.1
XP_016876664.1
9
3218
XM_017021176.1
XP_016876665.1
14
3045
XM_017021177.1
XP_016876666.1
14
3034
XM_017021178.1
XP_016876667.1
12
2872
XM_017021179.1
XP_016876668.1
11
2775
XM_017021180.1
XP_016876669.1
15
3087
XM_017021181.1
XP_016876670.1
13
2992
XM_017021182.1
XP_016876671.1
12
2925
XM_017021183.1
XP_016876672.1
10
2733
XM_017021184.1
XP_016876673.1
14
3045
XM_017021185.1
XP_016876674.1
7
3073
XM_017021186.1
XP_016876675.1
13
2992
XM_017021187.1
XP_016876676.1
11
2858
XM_017021188.1
XP_016876677.1
10
2733
XM_017021189.1
XP_016876678.1
12
2842
XM_017021190.1
XP_016876679.1
13
2992
XM_017021191.1
XP_016876680.1
11
2800
XM_017021192.1
XP_016876681.1
10
2733
XM_017021193.1
XP_016876682.1
10
2731
XM_017021194.1
XP_016876683.1
13
2992
XM_017021195.1
XP_016876684.1
13
2992
XM_017021196.1
XP_016876685.1
13
2992
XM_017021197.1
XP_016876686.1
12
1192
XM_017021198.1
XP_016876687.1
9
933
XM_017021199.1
XP_016876688.1
13
1234
XM_017021200.1
XP_016876689.1
10
1000
GenBank mRNA
AB007900.2
-
6
2208
AF090989.1
-
7
2471
AF090990.1
-
7
2471
AK122930.1
-
7
3073
AK302650.1
-
6
723
BC146777.1
-
6
2208
9007226
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
actin cytoskeleton reorganization
regulation of GTPase activity
ephrin receptor signaling pathway
regulation of synaptic plasticity
regulation of axonogenesis
regulation of small GTPase mediated signal transduction
regulation of dendritic spine morphogenesis
activation of GTPase activity
molecular_function
GTPase activator activity
ephrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0