Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015557.2 | NP_056372.1 | 19 | 3057 | ||
GenBank mRNA | AB007913.1 | - | 1 | 18 | ||
AF425231.1 | - | 19 | 3046 | |||
AK091386.1 | - | 10 | 1533 | |||
AK127046.1 | - | 19 | 2946 | |||
AL110281.1 | - | 8 | 1094 | |||
AL117491.1 | - | 9 | 1250 | |||
AL137693.1 | - | 8 | 1073 |