Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320766.1 | NP_001307695.1 | 12 | 2620 | ||
NM_015565.2 | NP_056380.2 | 13 | 2659 | |||
XM_006723987.3 | XP_006724050.1 | 13 | 2657 | |||
XM_017028316.1 | XP_016883805.1 | 12 | 2339 | |||
XM_017028317.1 | XP_016883806.1 | 3 | 934 | |||
GenBank mRNA | AB018257.2 | - | 13 | 2550 | ||
AL703566.1 | - | 1 | 42 | |||
BC140790.1 | - | 13 | 2572 | 9021658 | ||
BC150284.1 | - | 13 | 2550 | 8860138 |