Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320766.1 | NP_001307695.1 | 28 | 5283 | ||
NM_015565.2 | NP_056380.2 | 29 | 5322 | |||
XM_017028316.1 | XP_016883805.1 | 28 | 5002 | |||
XM_017028317.1 | XP_016883806.1 | 19 | 3597 | |||
GenBank mRNA | AB018257.2 | - | 29 | 5213 | ||
BC140790.1 | - | 29 | 5235 | 9021658 | ||
BC150284.1 | - | 29 | 5213 | 8860138 | ||
DB636695.1 | - | 2 | 260 |