Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015630.3 | NP_056445.3 | 14 | 2411 | ||
XM_011510941.2 | XP_011509243.1 | 14 | 2616 | |||
XM_011510943.2 | XP_011509245.1 | 13 | 2295 | |||
GenBank mRNA | AF286904.1 | - | 14 | 2403 | ||
AK001433.1 | - | 12 | 2002 | |||
AK002010.1 | - | 5 | 858 | |||
AK125789.1 | - | 15 | 2480 | |||
AL117558.1 | - | 3 | 405 | |||
BC093818.1 | - | 14 | 2323 | 7939663 | ||
BC093820.1 | - | 14 | 2323 | 7939665 |