Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018067.1 | NP_001018077.1 | 6 | 1026 | ||
NM_001018068.1 | NP_001018078.1 | 6 | 1008 | |||
NM_001018069.1 | NP_001018079.1 | 6 | 981 | |||
NM_015640.3 | NP_056455.3 | 6 | 963 | |||
GenBank mRNA | AF151813.1 | - | 6 | 940 | ||
AK074970.1 | - | 6 | 975 | |||
AK130606.1 | - | 6 | 909 | |||
AK222955.1 | - | 6 | 909 | |||
AK223138.1 | - | 6 | 909 | |||
AK312256.1 | - | 6 | 925 | |||
AL080119.1 | - | 6 | 908 | |||
AY032853.1 | - | 6 | 590 | |||
BC002488.2 | - | 6 | 935 | 3051198 | ||
BC003049.1 | - | 6 | 934 | 3547236 | ||
BC008045.2 | - | 6 | 957 | 3615828 | ||
BC017449.1 | - | 6 | 894 | 3535655 | ||
BC019273.1 | - | 6 | 963 | 2823760 | ||
BC020555.1 | - | 6 | 940 | 3446543 | ||
BC026916.1 | - | 6 | 958 | 4477452 | ||
BX647139.1 | - | 6 | 938 | |||
CR457383.1 | - | 6 | 823 |