Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SEPT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015129.5 | NP_055944.2 | 6, 7 | 1049 | ||
NM_145799.3 | NP_665798.1 | 6, 7 | 1049 | |||
NM_145800.3 | NP_665799.1 | 6, 7 | 1049 | |||
NM_145802.3 | NP_665801.1 | 6, 7 | 1049 | |||
XM_005262400.3 | XP_005262457.1 | 6, 7 | 857 | |||
XM_006724748.2 | XP_006724811.1 | 6, 7 | 857 | |||
XM_006724750.2 | XP_006724813.1 | 6, 7 | 857 | |||
XM_011531317.2 | XP_011529619.1 | 6, 7 | 857 | |||
XM_011531318.2 | XP_011529620.1 | 6, 7 | 857 | |||
GenBank mRNA | AF397023.1 | - | 6, 7 | 894 | ||
AF403058.1 | - | 6, 7 | 1049 | |||
AF403059.1 | - | 6, 7 | 1049 | |||
AF403060.1 | - | 6, 7 | 1049 | |||
AF403062.1 | - | 6, 7 | 1049 | |||
AK026589.1 | - | 6, 7 | 842 | |||
AK290977.1 | - | 6, 7 | 1101 | |||
AK298487.1 | - | 7, 8 | 956 | |||
AK299273.1 | - | 6, 7 | 904 | |||
AK303116.1 | - | 7, 8 | 926 | |||
AK303224.1 | - | 6, 7 | 972 | |||
AY005981.1 | - | 6, 7 | 941 | |||
AY034177.1 | - | 6, 7 | 1039 | |||
AY679521.1 | - | 6, 7 | 1049 | |||
BC009291.2 | - | 6, 7 | 833 | 4111673 | ||
BC011922.2 | - | 6, 7 | 837 | 4542179 | ||
BC036240.1 | - | 6, 7 | 876 | 5298284 | ||
BC069231.1 | - | 6, 7 | 821 | 4852319 | ||
D50918.1 | - | 777 |