Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FAM50B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012135.2 | NP_036267.1 | 2 | 853 | ||
XM_017010729.1 | XP_016866218.1 | 2 | 836 | |||
GenBank mRNA | BC001261.1 | - | 2 | 831 | 3457886 | |
Y18503.1 | - | 2 | 821 |