Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042483.1 | NP_001035948.1 | 3 | 707 | ||
NM_012385.2 | NP_036517.1 | 3 | 653 | |||
GenBank mRNA | AF069073.1 | - | 3 | 488 | ||
AF135266.2 | - | 3 | 444 | |||
BC002434.2 | - | 3 | 433 | 3347264 | ||
BF965595.1 | - | 3 | 488 |