Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014571.3 | NP_055386.1 | 2, 3 | 194 | ||
GenBank mRNA | AF311885.1 | - | 2, 3 | 143 | ||
AJ272215.1 | - | 2, 3 | 153 | |||
AK300463.1 | - | 2, 3 | 153 | |||
AL541019.3 | - | 2, 3 | 194 | |||
BC006087.1 | - | 2, 3 | 164 | 2964729 |