Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB1BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012278.2 | NP_036410.1 | 3 | 198 | ||
GenBank mRNA | AF110225.1 | - | 3 | 152 | ||
AF140690.1 | - | 3 | 168 | |||
BC108901.2 | - | 2 | 425 | 40004692 | ||
DA895605.1 | - | 3 | 201 |