Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3YL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282687.1 | NP_001269616.1 | 11 | 1094 | ||
NM_015677.3 | NP_056492.2 | 9 | 940 | |||
NR_104223.1 | 11 | 2499 | ||||
GenBank mRNA | AL050373.1 | - | 11 | 1107 | ||
BC008374.1 | - | 9 | 849 | 3996066 | ||
BC043234.1 | - | 11 | 2504 |