Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3YL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159597.2 | NP_001153069.1 | 6 | 602 | ||
NM_001282682.1 | NP_001269611.1 | 8 | 707 | |||
NM_001282687.1 | NP_001269616.1 | 8 | 756 | |||
NM_015677.3 | NP_056492.2 | 6 | 602 | |||
NR_104223.1 | 8 | 2161 | ||||
NR_104225.1 | 6 | 797 | ||||
NR_104226.1 | 8 | 1322 | ||||
NR_104227.1 | 8 | 1668 | ||||
GenBank mRNA | AK026507.1 | - | 6 | 537 | ||
AK026586.1 | - | 6 | 797 | |||
AK055801.1 | - | 6 | 1261 | |||
AK096440.1 | - | 8 | 1246 | |||
AK123829.1 | - | 9 | 2400 | |||
AK292312.1 | - | 8 | 651 | |||
AL050373.1 | - | 8 | 769 | |||
BC008374.1 | - | 6 | 511 | 3996066 | ||
BC008375.1 | - | 6 | 529 | 3996120 | ||
BC043234.1 | - | 8 | 2166 | |||
CR457361.1 | - | 5 | 489 |