Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014443.2 | NP_001014443.1 | 10 | 1633 | ||
NM_001319848.1 | NP_001306777.1 | 9 | 1492 | |||
NM_012475.4 | NP_036607.3 | 9 | 1492 | |||
XM_011509414.1 | XP_011507716.1 | 9 | 1496 | |||
XM_017001036.1 | XP_016856525.1 | 9 | 1555 | |||
XM_017001037.1 | XP_016856526.1 | 10 | 1609 | |||
XM_017001038.1 | XP_016856527.1 | 10 | 1645 | |||
XM_017001039.1 | XP_016856528.1 | 9 | 1498 | |||
XM_017001040.1 | XP_016856529.1 | 9 | 1555 | |||
XM_017001041.1 | XP_016856530.1 | 10 | 1294 | |||
XM_017001042.1 | XP_016856531.1 | 5 | 453 | |||
GenBank mRNA | AB208899.1 | - | 7 | 1904 | ||
AF177758.1 | - | 9 | 1343 | |||
AF217979.1 | - | 9 | 1416 | |||
AK292319.1 | - | 10 | 1578 | |||
AK310326.1 | - | 9 | 1525 | |||
AL157417.1 | - | 6 | 1525 | |||
BC003130.1 | - | 8 | 992 | |||
BC090946.1 | - | 10 | 1581 | 3877298 | ||
BC136291.1 | - | 8 | 1280 | 9020278 |