Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015693.3 | NP_056508.2 | 15, 16 | 2819 | ||
XM_011531844.2 | XP_011530146.1 | 15, 16 | 2843 | |||
XM_011531845.2 | XP_011530147.1 | 14, 15 | 2792 | |||
XM_011531846.1 | XP_011530148.1 | 15, 16 | 2791 | |||
XM_011531847.1 | XP_011530149.1 | 15, 16 | 2228 | |||
XM_011531848.2 | XP_011530150.1 | 10, 11 | 1654 | |||
XM_017008025.1 | XP_016863514.1 | 12, 13 | 1762 | |||
GenBank mRNA | AB033110.1 | - | 15, 16 | 2786 | ||
AK123320.1 | - | 8, 9 | 2159 | |||
AK291481.1 | - | 15, 16 | 2801 | |||
BC051698.1 | - | 17, 18 | 5287 | |||
BC130611.1 | - | 15, 16 | 2785 | 40146642 |