Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164623.1 | NP_001158095.1 | 9 | 1174 | ||
NM_012154.3 | NP_036286.2 | 9 | 1174 | |||
XM_011516965.2 | XP_011515267.1 | 9 | 4250 | |||
XM_011516966.2 | XP_011515268.1 | 9 | 1991 | |||
XM_011516968.2 | XP_011515270.2 | 9 | 1325 | |||
XM_017013317.1 | XP_016868806.1 | 8 | 943 | |||
GenBank mRNA | BC007633.1 | - | 1 | 58 | ||
BC018727.1 | - | 4 | 421 | |||
BC125213.1 | - | 8 | 980 | |||
BT007229.1 | - | 3 | 311 |