Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280796.1 | NP_001267725.1 | 7, 8 | 1141 | ||
NM_001280797.1 | NP_001267726.1 | 6, 7 | 1011 | |||
NM_001280798.1 | NP_001267727.1 | 4, 5 | 789 | |||
NM_013267.3 | NP_037399.2 | 7, 8 | 1141 | |||
XM_005268797.1 | XP_005268854.1 | 7, 8 | 768 | |||
XM_017019180.1 | XP_016874669.1 | 6, 7 | 638 | |||
GenBank mRNA | AF110330.1 | - | 7, 8 | 896 | ||
AF223944.1 | - | 7, 8 | 1068 | |||
AF279698.1 | - | 4, 5 | 395 | |||
AK000467.1 | - | 7, 8 | 888 | |||
AK294822.1 | - | 7, 8 | 1086 | |||
AK303772.1 | - | 7, 8 | 942 | |||
BC025661.1 | - | 6, 7 | 726 | |||
DA060302.1 | - | 4, 5 | 373 | |||
HM775422.1 | - | 6, 7 | 760 | |||
HM775423.1 | - | 5, 6 | 630 | |||
HM775424.1 | - | 6, 7 | 760 |