Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198558.1 | NP_001185487.1 | 1 | 147 | ||
NM_014441.2 | NP_055256.1 | 1 | 147 | |||
XM_006723146.3 | XP_006723209.1 | 1 | 119 | |||
XM_011526730.2 | XP_011525032.1 | 1 | 119 | |||
XM_011526732.2 | XP_011525034.1 | 1 | 119 | |||
XM_017026595.1 | XP_016882084.1 | 1 | 119 | |||
XM_017026596.1 | XP_016882085.1 | 1 | 119 | |||
GenBank mRNA | AB026265.1 | - | 1 | 98 | ||
AF227924.1 | - | 1 | 119 | |||
AF247180.1 | - | 1 | 63 | |||
AK297043.1 | - | 1 | 119 | |||
AK315272.1 | - | 1 | 90 | |||
AY358913.1 | - | 1 | 94 | |||
BC035365.1 | - | 1 | 101 | 5176457 | ||
BP305376.1 | - | 1 | 148 |