Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014482.1 | NP_055297.1 | 2 | 1264 | ||
GenBank mRNA | AF101441.1 | - | 2 | 1264 | ||
BC069080.1 | - | 2 | 1118 | 7216894 | ||
BC101734.1 | - | 2 | 1128 | 8069240 | ||
BC105063.1 | - | 2 | 1128 | 8144066 |