Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PGAP2 SIRNA Assays ›
Gene Symbol:
PGAP2
Gene Name:
post-GPI attachment to proteins 2
Gene Aliases:
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
Chromosome Location:
Chr.11: 3797724 - 3826371 on Build GRCh38
UniGene ID:
Hs.133968
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s26139
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PGAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145438.2
NP_001138910.1
4
581
NM_001256235.1
NP_001243164.1
5
522
NM_001256236.1
NP_001243165.1
5
764
NM_001256237.1
NP_001243166.1
4
581
NM_001256238.1
NP_001243167.1
4
495
NM_001256239.1
NP_001243168.1
3
511
NM_001256240.1
NP_001243169.1
3
511
NM_001283038.1
NP_001269967.1
4
581
NM_001283039.1
NP_001269968.1
4
534
NM_014489.3
NP_055304.1
4
694
NR_027016.2
2
336
NR_027017.2
5
644
NR_027018.2
5
533
NR_045923.1
3
406
NR_045927.1
3
406
NR_045929.1
3
464
NR_104270.1
3
406
NR_104271.1
5
666
NR_104272.1
2
336
XM_006718181.3
XP_006718244.1
3
1172
XM_006718183.3
XP_006718246.1
5
589
XM_006718185.2
XP_006718248.1
4
481
XM_006718186.2
XP_006718249.1
5
641
XM_006718190.3
XP_006718253.1
4
797
XM_006718191.3
XP_006718254.1
4
797
XM_006718192.1
XP_006718255.1
6
715
XM_006718193.2
XP_006718256.1
5
522
XM_011519990.2
XP_011518292.2
5
1048
XM_011519991.2
XP_011518293.2
5
1051
XM_011519992.1
XP_011518294.1
6
820
XM_011519993.2
XP_011518295.2
5
1067
XM_011519994.2
XP_011518296.2
5
1067
XM_011519995.2
XP_011518297.1
5
971
XM_011519996.1
XP_011518298.1
6
772
XM_011519997.1
XP_011518299.1
6
1113
XM_011519998.2
XP_011518300.1
6
791
XM_011519999.1
XP_011518301.1
6
750
XM_011520000.1
XP_011518302.1
5
664
XM_011520001.1
XP_011518303.1
4
456
XM_011520002.1
XP_011518304.1
4
456
XM_011520003.2
XP_011518305.1
5
980
XM_011520004.2
XP_011518306.1
5
655
XM_011520005.2
XP_011518307.1
4
552
XM_011520006.2
XP_011518308.1
5
936
XM_011520007.2
XP_011518309.1
6
705
XM_017017556.1
XP_016873045.1
4
456
XM_017017557.1
XP_016873046.1
4
604
XM_017017558.1
XP_016873047.1
4
778
XM_017017559.1
XP_016873048.1
5
641
XM_017017560.1
XP_016873049.1
5
641
GenBank mRNA
AF159615.1
-
4
696
AK292181.1
-
5
658
AK295202.1
-
5
533
AK301991.1
-
5
614
AK307359.1
-
3
323
AK308525.1
-
3
317
AL049261.1
-
2
946
AL096753.1
-
5
522
AY373030.1
-
2
385
BC009930.2
-
4
468
3506614
BG707484.1
-
3
300
BI460657.1
-
6
778
BI759334.1
-
3
403
BI768071.1
-
3
468
BI771208.1
-
2
297
BI772237.1
-
4
474
BI911526.1
-
5
555
BX340489.2
-
3
448
BX356273.2
-
4
488
BX379511.2
-
4
463
BX448155.2
-
4
440
CB988453.1
-
2
363
CD675009.1
-
1
47
CD675113.1
-
3
299
CX872317.1
-
2
264
KU178608.1
-
2
385
KU178609.1
-
3
470
KU178610.1
-
3
470
KU178611.1
-
2
385
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
protein transport
signal transduction in response to DNA damage
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
protein binding
protein transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-tbml7:80/10.239.166.229:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0