Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014972.2 | NP_001014972.1 | 20, 21 | 3571 | ||
NM_001252612.1 | NP_001239541.1 | 20, 21 | 3897 | |||
NM_001252613.1 | NP_001239542.1 | 20, 21 | 3581 | |||
NM_014497.4 | NP_055312.2 | 20, 21 | 3581 | |||
XM_005264263.1 | XP_005264320.1 | 19, 20 | 2045 | |||
XM_006711989.1 | XP_006712052.1 | 19, 20 | 2057 | |||
XM_017003809.1 | XP_016859298.1 | 20, 21 | 3672 | |||
XM_017003811.1 | XP_016859300.1 | 20, 21 | 3667 | |||
GenBank mRNA | AF273049.1 | - | 10, 11 | 1014 | ||
AF534078.1 | - | 19, 20 | 3261 | |||
AK291198.1 | - | 20, 21 | 3545 | |||
AK303723.1 | - | 18, 19 | 2041 | |||
BC083513.1 | - | 20, 21 | 3540 | 5540321 | ||
BC143728.1 | - | 20, 21 | 3534 | 9052242 | ||
BX537425.1 | - | 20, 21 | 3545 | |||
CR749322.1 | - | 20, 21 | 3894 | |||
D83032.1 | - | 20, 21 | 3573 | |||
DB130984.1 | - | 4, 5 | 422 |