Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POLL SIRNA Assays ›
Gene Symbol:
POLL
Gene Name:
polymerase (DNA) lambda
Gene Aliases:
BETAN, POLKAPPA
Chromosome Location:
Chr.10: 101578882 - 101588270 on Build GRCh38
UniGene ID:
Hs.523230
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s26196
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POLL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001174084.1
NP_001167555.1
3
673
NM_001174085.1
NP_001167556.1
3
610
NM_013274.3
NP_037406.1
3
610
NR_033406.1
3
209
XM_011539650.1
XP_011537952.1
3
278
XM_011539651.1
XP_011537953.1
3
278
XM_011539652.1
XP_011537954.1
3
566
XM_011539653.1
XP_011537955.1
3
559
XM_011539654.1
XP_011537956.1
3
278
XM_011539655.1
XP_011537957.1
3
278
XM_011539656.1
XP_011537958.1
3
278
XM_011539657.1
XP_011537959.1
3
505
XM_011539667.1
XP_011537969.1
3
240
XM_017016084.1
XP_016871573.1
3
278
XM_017016085.1
XP_016871574.1
3
278
XM_017016086.1
XP_016871575.1
2
986
XM_017016087.1
XP_016871576.1
3
278
XM_017016088.1
XP_016871577.1
3
278
XM_017016090.1
XP_016871579.1
3
278
XM_017016092.1
XP_016871581.1
3
559
GenBank mRNA
AF161019.1
-
2
239
AF283478.1
-
2
178
AJ131890.1
-
3
549
AK021600.1
-
3
281
AK022476.1
-
3
187
AK292225.1
-
3
532
AK293421.1
-
3
270
AK293657.1
-
2
115
AY302442.1
-
2
178
BC068529.1
-
3
562
5267859
DA107389.1
-
3
209
DC379162.1
-
3
209
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-directed DNA polymerase
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication
nucleotide-excision repair
double-strand break repair via nonhomologous end joining
somatic hypermutation of immunoglobulin genes
DNA biosynthetic process
DNA binding
DNA-directed DNA polymerase activity
lyase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-462rt:80/10.239.167.23:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0