Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POLL SIRNA Assays ›
Gene Symbol:
POLL
Gene Name:
polymerase (DNA) lambda
Gene Aliases:
BETAN, POLKAPPA
Chromosome Location:
Chr.10: 101578882 - 101588270 on Build GRCh38
UniGene ID:
Hs.523230
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s26198
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POLL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001174084.1
NP_001167555.1
2
568
NM_001174085.1
NP_001167556.1
2
505
NM_001308382.1
NP_001295311.1
2
186
NM_013274.3
NP_037406.1
2
505
XM_006717775.1
XP_006717838.1
1
907
XM_006717776.1
XP_006717839.1
2
135
XM_006717777.1
XP_006717840.1
1
907
XM_011539650.1
XP_011537952.1
2
173
XM_011539651.1
XP_011537953.1
2
173
XM_011539652.1
XP_011537954.1
2
461
XM_011539653.1
XP_011537955.1
2
454
XM_011539654.1
XP_011537956.1
2
173
XM_011539655.1
XP_011537957.1
2
173
XM_011539656.1
XP_011537958.1
2
173
XM_011539659.1
XP_011537961.1
1
907
XM_011539660.1
XP_011537962.1
2
163
XM_011539662.1
XP_011537964.1
1
907
XM_011539663.1
XP_011537965.1
2
135
XM_011539664.1
XP_011537966.1
1
907
XM_011539665.2
XP_011537967.1
2
495
XM_011539666.1
XP_011537968.1
2
173
XM_011539667.1
XP_011537969.1
2
135
XM_017016084.1
XP_016871573.1
2
173
XM_017016085.1
XP_016871574.1
2
173
XM_017016086.1
XP_016871575.1
1
881
XM_017016087.1
XP_016871576.1
2
173
XM_017016088.1
XP_016871577.1
2
173
XM_017016089.1
XP_016871578.1
2
436
XM_017016090.1
XP_016871579.1
2
173
XM_017016091.1
XP_016871580.1
1
907
XM_017016092.1
XP_016871581.1
2
454
XM_017016093.1
XP_016871582.1
2
432
GenBank mRNA
AF161019.1
-
1
134
AF283478.1
-
1
73
AJ131890.1
-
2
444
AK021600.1
-
2
176
AK292225.1
-
2
427
AK293421.1
-
2
165
AK293602.1
-
2
135
AK300862.1
-
2
165
AY302442.1
-
1
73
BC003548.1
-
1
839
3536366
BC068529.1
-
2
457
5267859
BI906881.1
-
2
146
DC409528.1
-
2
492
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00203191_m1
Panther Classification:
Molecular Function -
DNA-directed DNA polymerase
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication
nucleotide-excision repair
double-strand break repair via nonhomologous end joining
somatic hypermutation of immunoglobulin genes
DNA biosynthetic process
DNA binding
DNA-directed DNA polymerase activity
lyase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-cq5mm:80/10.239.166.46:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0