Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014510.2 | NP_055325.2 | 3 | 2410 | ||
NM_033026.5 | NP_149015.2 | 3 | 2410 | |||
XM_017012006.1 | XP_016867495.1 | 3 | 3685 | |||
XM_017012007.1 | XP_016867496.1 | 3 | 3669 | |||
GenBank mRNA | AK297315.1 | - | 3 | 2413 | ||
Y19188.1 | - | 5 | 1851 |