Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013239.4 | NP_037371.2 | 13 | 2094 | ||
GenBank mRNA | AB209137.1 | - | 8 | 5788 | ||
AF135016.1 | - | 12 | 1458 | |||
AF155098.1 | - | 7 | 869 | |||
AL157501.1 | - | 5 | 805 | |||
BC009032.1 | - | 5 | 1051 | |||
BC011180.1 | - | 7 | 871 | |||
BC063429.1 | - | 13 | 1849 | 5239974 | ||
CB218994.1 | - | 2 | 287 |