Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011527533.2 | XP_011525835.1 | 2 | 568 | ||
XM_011527534.2 | XP_011525836.1 | 2 | 571 | |||
XM_011527535.2 | XP_011525837.1 | 2 | 515 | |||
XM_011527536.2 | XP_011525838.1 | 2 | 407 | |||
XM_011527537.2 | XP_011525839.1 | 2 | 407 | |||
XM_011527538.2 | XP_011525840.1 | 2 | 570 | |||
XM_011527539.2 | XP_011525841.1 | 2 | 569 | |||
XM_017027511.1 | XP_016883000.1 | 2 | 571 | |||
GenBank mRNA | AK300170.1 | - | 2 | 408 | ||
AK300182.1 | - | 2 | 506 | |||
AK308174.1 | - | 2 | 515 | |||
AK309903.1 | - | 2 | 407 | |||
AU138048.1 | - | 2 | 515 | |||
BC035359.1 | - | 2 | 429 | 5176194 | ||
BC140798.1 | - | 2 | 379 | 9021666 | ||
CA495365.1 | - | 1 | 114 | |||
D86358.1 | - | 2 | 563 | |||
D86359.1 | - | 2 | 563 | |||
U71382.1 | - | 2 | 363 |