Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORMDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014182.4 | NP_054901.1 | 3 | 408 | ||
GenBank mRNA | AB064961.1 | - | 3 | 362 | ||
AF161509.1 | - | 3 | 369 | |||
AF173375.1 | - | 3 | 378 | |||
AF395705.1 | - | 2 | 323 | |||
AF395706.1 | - | 2 | 328 | |||
AF395707.1 | - | 3 | 505 | |||
AK223180.1 | - | 3 | 349 | |||
AK314046.1 | - | 3 | 389 | |||
BC012543.2 | - | 2 | 318 | 4272865 |