Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013242.2 | NP_037374.1 | 3 | 510 | ||
NR_136632.1 | 3 | 543 | ||||
GenBank mRNA | AF087845.1 | - | 3 | 490 | ||
AF093680.1 | - | 3 | 489 | |||
AK027099.1 | - | 3 | 431 | |||
AK297819.1 | - | 3 | 465 | |||
BC005152.2 | - | 3 | 479 | 3528301 | ||
CN364433.1 | - | 3 | 470 |