Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114094.1 | NP_001107566.1 | 15 | 1217 | ||
NM_013314.3 | NP_037446.1 | 16 | 1286 | |||
NR_047680.1 | 15 | 1050 | ||||
NR_047681.1 | 14 | 998 | ||||
NR_047682.1 | 14 | 993 | ||||
XM_011539728.2 | XP_011538030.1 | 16 | 1163 | |||
XM_017016159.1 | XP_016871648.1 | 15 | 1095 | |||
GenBank mRNA | AF068180.1 | - | 16 | 1261 | ||
AF068181.1 | - | 15 | 1192 | |||
AK027779.1 | - | 16 | 1221 | |||
AK225546.1 | - | 15 | 1200 | |||
AM180333.1 | - | 15 | 1039 | |||
AM180334.1 | - | 15 | 1039 | |||
AM180335.1 | - | 12 | 677 | |||
AM180338.1 | - | 14 | 880 | |||
AM180339.1 | - | 14 | 875 | |||
AM180343.1 | - | 15 | 1058 | |||
AM180344.1 | - | 13 | 948 | |||
AM180346.1 | - | 13 | 975 | |||
BC018906.2 | - | 16 | 1200 | 4053894 | ||
BC110298.1 | - | 15 | 1042 | |||
BF975580.1 | - | 4 | 217 |